Learn More
MSC Vermont Gage Pin Gage, 6.63mm Diameter, Class ZZ, Minus, Tool Steel
Catalog No. 5000527861
Encompass
This Vermont Gage Pin Gage is a precision measuring tool for accurate hole size measurement. Crafted from durable tool steel (Rc 60/65), it offers longevity and consistent performance. With a 6.63mm diameter and Class ZZ designation, it features a tight tolerance of +.000/-.005. It includes a certificate of accuracy with NIST traceability, ensuring calibration and adherence to standards.
  • Durable construction: tool steel (Rc 60/65) for long-lasting use.
  • High precision: Class ZZ with +.000/-.005 tolerance for accurate hole measurements.
  • Quality finish: steel with a 10 microfinish or better.
  • NIST traceability: certificate of accuracy included for calibration and standards compliance.
  • Application: measures hole sizes within +.000/-.005".
Catalog No. 50-005-27861 Supplier MSC Supplier No. 40434086
This Vermont Gage Pin Gage is a precision measuring tool for accurate hole size measurement. Crafted from durable tool steel (Rc 60/65), it offers longevity and consistent performance. With a 6.63mm diameter and Class ZZ designation, it features a tight tolerance of +.000/-.005. It includes a certificate of accuracy with NIST traceability, ensuring calibration and adherence to standards.
  • Durable construction: tool steel (Rc 60/65) for long-lasting use.
  • High precision: Class ZZ with +.000/-.005 tolerance for accurate hole measurements.
  • Quality finish: steel with a 10 microfinish or better.
  • NIST traceability: certificate of accuracy included for calibration and standards compliance.
  • Application: measures hole sizes within +.000/-.005".