Learn More
MSC Vermont Gage Pin Gage, 6.81 mm, Class ZZ, Steel, Plus Tolerance
Catalog No. 5000528061
Encompass
Vermont Gage pin gages are designed for precise measurement applications. This particular pin gage is a Class ZZ, plus tolerance type, made from durable steel, with a diameter of 6.81 mm. It features a reversible design and is part of a metric system, suitable for various inspection tasks.
  • Class ZZ
  • Plus tolerance
  • Steel construction
  • 6.81 mm diameter
  • Metric system of measurement
  • Reversible style
  • Member only type
  • Manufactured in the US
Catalog No. 50-005-28061 Supplier MSC Supplier No. 68097864
Vermont Gage pin gages are designed for precise measurement applications. This particular pin gage is a Class ZZ, plus tolerance type, made from durable steel, with a diameter of 6.81 mm. It features a reversible design and is part of a metric system, suitable for various inspection tasks.
  • Class ZZ
  • Plus tolerance
  • Steel construction
  • 6.81 mm diameter
  • Metric system of measurement
  • Reversible style
  • Member only type
  • Manufactured in the US