Learn More
MSC Vermont Gage Pin Gage, 6.84 mm, Class ZZ Minus, Tool Steel, 2 in
Catalog No. 5000532730
Encompass
This Vermont Gage Class ZZ Minus Pin Gage is a precision gaging tool designed for critical measurements. It features a 6.84 mm diameter and is constructed from durable tool steel with a 60/65 Rc hardness. The gage offers a Class ZZ minus tolerance, ensuring high accuracy for various applications.
  • Measures hole diameters.
  • Calculates distances between holes.
  • Gages slot widths and locations.
  • Used for Go/NoGo inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Features a 10 microinch finish or better.
  • Class ZZ (.0002 in or 0.005 mm) tolerance.
  • 2 in length.
  • Round within .0001 in or 0.0025 mm.
Catalog No. 50-005-32730 Supplier MSC Supplier No. 40434292
This Vermont Gage Class ZZ Minus Pin Gage is a precision gaging tool designed for critical measurements. It features a 6.84 mm diameter and is constructed from durable tool steel with a 60/65 Rc hardness. The gage offers a Class ZZ minus tolerance, ensuring high accuracy for various applications.
  • Measures hole diameters.
  • Calculates distances between holes.
  • Gages slot widths and locations.
  • Used for Go/NoGo inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Features a 10 microinch finish or better.
  • Class ZZ (.0002 in or 0.005 mm) tolerance.
  • 2 in length.
  • Round within .0001 in or 0.0025 mm.