Learn More
MSC Vermont Gage Pin Gage, .6855 in, Class ZZ, Minus, Black Guard
Catalog No. 5000531027
Encompass
This precision pin gage is designed for accurate measurement and inspection in demanding gaging applications. It features a a .6855 inch diameter with a Class ZZ tolerance, ensuring reliable performance and precision. The Black Guard material provides durability and corrosion resistance.
  • Provides accurate measurement and inspection
  • Features a .6855 inch diameter
  • Has a 2 inch overall length
  • Made from Black Guard material for durability
  • Offers Class ZZ tolerance
  • Has a tolerance of 0.0002 inch
  • Designed as a minus type gage for "NoGo" applications
Catalog No. 50-005-31027 Supplier MSC Supplier No. 72026222
This precision pin gage is designed for accurate measurement and inspection in demanding gaging applications. It features a a .6855 inch diameter with a Class ZZ tolerance, ensuring reliable performance and precision. The Black Guard material provides durability and corrosion resistance.
  • Provides accurate measurement and inspection
  • Features a .6855 inch diameter
  • Has a 2 inch overall length
  • Made from Black Guard material for durability
  • Offers Class ZZ tolerance
  • Has a tolerance of 0.0002 inch
  • Designed as a minus type gage for "NoGo" applications