Learn More
MSC Vermont Gage Pin Gage, Class X, 0.3175 Inch, Go Plus, Tool Steel
Catalog No. 5000534383
Encompass
This Class X Go Plus pin gage is designed for precise measurement and calibration tasks. It serves as an accurate, cost-effective standard for calibrating measuring equipment or directly gaging parts, making it suitable for various shop floor applications.
  • Class X tolerance for high precision.
  • Go (Plus) type for specific measurement applications.
  • NIST traceable, ensuring accuracy and reliability.
  • Constructed from durable tool steel with 60/65 Rc (cold stabilized) hardness.
  • Measures 2 inch in length.
  • Features a 0.3175 inch diameter.
  • Maintains a tight tolerance of 0.00004 "
  • Finished with a 4 micro coating.
Catalog No. 50-005-34383 Supplier MSC Supplier No. 88731757
This Class X Go Plus pin gage is designed for precise measurement and calibration tasks. It serves as an accurate, cost-effective standard for calibrating measuring equipment or directly gaging parts, making it suitable for various shop floor applications.
  • Class X tolerance for high precision.
  • Go (Plus) type for specific measurement applications.
  • NIST traceable, ensuring accuracy and reliability.
  • Constructed from durable tool steel with 60/65 Rc (cold stabilized) hardness.
  • Measures 2 inch in length.
  • Features a 0.3175 inch diameter.
  • Maintains a tight tolerance of 0.00004 "
  • Finished with a 4 micro coating.