Learn More
MSC Vermont Gage Pin Gage, Class X, 0.3747 in, 2 in, 4 Micro Coated
Catalog No. 5000536197
Encompass
The Vermont Gage Class X Plus Pin Gage is designed for precise measurement and calibration in demanding applications. This gage features a 0.3747 inch diameter and a 2 inch length, with a 4 micro coated finish for enhanced durability and accuracy.
  • 4 micro coated finish.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Features a precise 0.00004 inch tolerance.
Catalog No. 50-005-36197 Supplier MSC Supplier No. 80297096
The Vermont Gage Class X Plus Pin Gage is designed for precise measurement and calibration in demanding applications. This gage features a 0.3747 inch diameter and a 2 inch length, with a 4 micro coated finish for enhanced durability and accuracy.
  • 4 micro coated finish.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Features a precise 0.00004 inch tolerance.