Learn More
MSC Vermont Gage Pin Gage, Class X, Go Tolerance, 0.2940 Inch Diameter, Tool Steel
Catalog No. 5000516408
Encompass
These Class X standard pin gages are designed for precise measurement, serving as masters for equipment calibration or for direct part gaging. They offer accurate and cost-effective measurement standards suitable for both shop floor and metrology laboratory applications.
  • Certificate of accuracy included
  • ISO 17025 accredited and non-accredited calibration certificates available for purchase
  • Manufactured from tool steel with 60/65 Rc hardness
  • Features a 4 microinch finish or better
  • Roundness within 1/2 the diameter tolerance
Catalog No. 50-005-16408 Supplier MSC Supplier No. 88729405
These Class X standard pin gages are designed for precise measurement, serving as masters for equipment calibration or for direct part gaging. They offer accurate and cost-effective measurement standards suitable for both shop floor and metrology laboratory applications.
  • Certificate of accuracy included
  • ISO 17025 accredited and non-accredited calibration certificates available for purchase
  • Manufactured from tool steel with 60/65 Rc hardness
  • Features a 4 microinch finish or better
  • Roundness within 1/2 the diameter tolerance