Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.0240 in Diameter, 2 in Length
Catalog No. 5000528866
Encompass
This pin gage is a precision measurement tool designed for accurate calibration of measuring equipment or direct gaging of parts. Made from durable tool steel, it features a 0.0240 inch diameter and a 2 inch length, with a Class X Minus tolerance for high accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Applicable for calibration of measuring equipment or direct gaging of parts.
  • Features a 4 micro coating/finish.
  • Manufactured in the United States.
Catalog No. 50-005-28866 Supplier MSC Supplier No. 88802400
This pin gage is a precision measurement tool designed for accurate calibration of measuring equipment or direct gaging of parts. Made from durable tool steel, it features a 0.0240 inch diameter and a 2 inch length, with a Class X Minus tolerance for high accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Applicable for calibration of measuring equipment or direct gaging of parts.
  • Features a 4 micro coating/finish.
  • Manufactured in the United States.