Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.0295 inch Diameter, 2 inch Long, Tool Steel
Catalog No. 5000532076
Encompass
This Vermont Gage pin gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. Made from durable tool steel, it features a 0.0295 inch diameter and a 2 inch length, ensuring accuracy and reliability in demanding environments.
  • 4 micro coated
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-32076 Supplier MSC Supplier No. 88802954
This Vermont Gage pin gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. Made from durable tool steel, it features a 0.0295 inch diameter and a 2 inch length, ensuring accuracy and reliability in demanding environments.
  • 4 micro coated
  • Includes certificate of accuracy with NIST traceability