Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.0410 inch diameter, 2 inch long
Catalog No. 5000527559
Encompass
This pin gage is designed for precise calibration of measuring equipment or for direct gaging of parts. It ensures accuracy with its Class X Minus tolerance and comes with a Certificate of Accuracy that offers NIST traceability.
  • Diameter of 0.0410 inch
  • Tolerance of -0.00004 inch
  • Constructed from tool steel
  • Used for calibrating measuring equipment or gaging parts directly
  • Includes a certificate of accuracy with NIST traceability
  • Length of 2 inch
  • Features a 4 micro coating
Catalog No. 50-005-27559 Supplier MSC Supplier No. 88804109
This pin gage is designed for precise calibration of measuring equipment or for direct gaging of parts. It ensures accuracy with its Class X Minus tolerance and comes with a Certificate of Accuracy that offers NIST traceability.
  • Diameter of 0.0410 inch
  • Tolerance of -0.00004 inch
  • Constructed from tool steel
  • Used for calibrating measuring equipment or gaging parts directly
  • Includes a certificate of accuracy with NIST traceability
  • Length of 2 inch
  • Features a 4 micro coating