Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.0628 in Diameter, 2 in Length
Catalog No. 5000531392
Encompass
The Class X Minus pin gage is designed for precise calibration of measuring equipment or direct part gaging. It features a 0.0628 inch diameter and a 2 inch length, crafted from durable tool steel, and coated with a 4 micro finish. This gage ensures high accuracy with a tolerance of -0.00004 inch and comes with a certificate of accuracy traceable to NIST.
  • Tolerance of -0.00004 inch for high precision.
  • Constructed from durable tool steel for longevity.
  • Suitable for calibrating measuring equipment or directly gaging parts.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coating/finish.
Catalog No. 50-005-31392 Supplier MSC Supplier No. 80340771
The Class X Minus pin gage is designed for precise calibration of measuring equipment or direct part gaging. It features a 0.0628 inch diameter and a 2 inch length, crafted from durable tool steel, and coated with a 4 micro finish. This gage ensures high accuracy with a tolerance of -0.00004 inch and comes with a certificate of accuracy traceable to NIST.
  • Tolerance of -0.00004 inch for high precision.
  • Constructed from durable tool steel for longevity.
  • Suitable for calibrating measuring equipment or directly gaging parts.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coating/finish.