Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.108" Dia, 2" Long, 4 Micro Finish
Catalog No. 5000510467
Encompass
This pin gage is designed for precise measurement applications. It features a 0.108 inch diameter and a 2 inch length, with a 4 micro finish, making it suitable for calibrating equipment or directly gaging parts. It also includes NIST traceability.
  • For calibration of measuring equipment or to gage parts directly
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Diameter of 0.1080 inch
  • Length of 2 inches
  • Features a 4 micro finish
  • Class X minus tolerance of -0.00004 inch
Catalog No. 50-005-10467 Supplier MSC Supplier No. 88810809
This pin gage is designed for precise measurement applications. It features a 0.108 inch diameter and a 2 inch length, with a 4 micro finish, making it suitable for calibrating equipment or directly gaging parts. It also includes NIST traceability.
  • For calibration of measuring equipment or to gage parts directly
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Diameter of 0.1080 inch
  • Length of 2 inches
  • Features a 4 micro finish
  • Class X minus tolerance of -0.00004 inch