Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.1290 in Diameter, 2 in Long, Tool Steel
Catalog No. 5000531332
Encompass
The Vermont Gage Class X Minus Pin Gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. Made from durable tool steel, this pin gage ensures reliable performance and comes with a Certificate of Accuracy for NIST traceability.
  • Class X Minus tolerance
  • 0.1290 inch diameter
  • 2 inch length
  • Made from tool steel
  • Suitable for calibration of measuring equipment
  • Can be used for direct part gaging
  • Includes certificate of accuracy with NIST traceability
  • Features a 4 micro finish
Catalog No. 50-005-31332 Supplier MSC Supplier No. 88812904
The Vermont Gage Class X Minus Pin Gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. Made from durable tool steel, this pin gage ensures reliable performance and comes with a Certificate of Accuracy for NIST traceability.
  • Class X Minus tolerance
  • 0.1290 inch diameter
  • 2 inch length
  • Made from tool steel
  • Suitable for calibration of measuring equipment
  • Can be used for direct part gaging
  • Includes certificate of accuracy with NIST traceability
  • Features a 4 micro finish