Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.1600 in Diameter, 2 in Length, 4 Micro Finish
Catalog No. 5000514417
Encompass
This pin gage is designed for precise measurement and calibration tasks. It features a 0.1600 in diameter and a 2 in length, with a 4 Micro finish, making it suitable for demanding laboratory and industrial applications.
  • Includes a Certificate of Accuracy with NIST Traceability
  • Designed for calibration of measuring equipment or to gage parts directly
  • Made from durable tool steel
  • Features a 4 micro finish for enhanced precision
Catalog No. 50-005-14417 Supplier MSC Supplier No. 88816004
This pin gage is designed for precise measurement and calibration tasks. It features a 0.1600 in diameter and a 2 in length, with a 4 Micro finish, making it suitable for demanding laboratory and industrial applications.
  • Includes a Certificate of Accuracy with NIST Traceability
  • Designed for calibration of measuring equipment or to gage parts directly
  • Made from durable tool steel
  • Features a 4 micro finish for enhanced precision