Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.1710 Inch, NoGo
Catalog No. 5000529036
Encompass
This Class X Minus Pin Gage is designed for precise measurement applications in both shop floor and metrology laboratory settings. It can be used as a master to calibrate measuring equipment or to directly gage parts. Made from durable tool steel, this NoGo type pin gage features a 0.1710 inch diameter and a high-quality finish, ensuring accuracy and reliability.
  • Made from tool steel with 60/65 Rc hardness for durability.
  • Features a 0.1710 inch diameter for accurate measurements.
  • Class X minus tolerance of -0.00004 "
  • Provides a 4 microinch finish or better for precise contact.
  • Overall length of 2 inches.
  • Suitable for calibrating measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-29036 Supplier MSC Supplier No. 88817101
This Class X Minus Pin Gage is designed for precise measurement applications in both shop floor and metrology laboratory settings. It can be used as a master to calibrate measuring equipment or to directly gage parts. Made from durable tool steel, this NoGo type pin gage features a 0.1710 inch diameter and a high-quality finish, ensuring accuracy and reliability.
  • Made from tool steel with 60/65 Rc hardness for durability.
  • Features a 0.1710 inch diameter for accurate measurements.
  • Class X minus tolerance of -0.00004 "
  • Provides a 4 microinch finish or better for precise contact.
  • Overall length of 2 inches.
  • Suitable for calibrating measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.