Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.1870 Inch Diameter, 2 Inch Length, 4 Micro Coated
Catalog No. 5000530841
Encompass
This Class X Minus Pin Gage is designed for the calibration of measuring equipment or for directly gaging parts. It is constructed from durable tool steel and features a 4 micro coating/finish, ensuring precision for various applications.
  • Designed for calibration of measuring equipment or directly gaging parts
  • Constructed from tool steel
  • Features a 4 micro coating/finish
  • Diameter of 0.1870 inch
  • Length of 2 inch
  • Tolerance of -0.00004 inch
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-30841 Supplier MSC Supplier No. 88818703
This Class X Minus Pin Gage is designed for the calibration of measuring equipment or for directly gaging parts. It is constructed from durable tool steel and features a 4 micro coating/finish, ensuring precision for various applications.
  • Designed for calibration of measuring equipment or directly gaging parts
  • Constructed from tool steel
  • Features a 4 micro coating/finish
  • Diameter of 0.1870 inch
  • Length of 2 inch
  • Tolerance of -0.00004 inch
  • Includes a certificate of accuracy with NIST traceability