Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.1880 in Diameter, 2 in Long
Catalog No. 5000515894
Encompass
This pin gage is a Class X Minus instrument designed for precise measurement applications, ideal for calibrating measuring equipment or directly gauging parts. It features a diameter of 0.1880 inches and a length of 2 inches, constructed from durable tool steel with a 4 Micro coating for enhanced performance.
  • Includes certificate of accuracy with NIST traceability
  • 4 Micro coated finish
  • Manufactured from tool steel
  • Suitable for calibration of measuring equipment or direct part gauging
Catalog No. 50-005-15894 Supplier MSC Supplier No. 88818802
This pin gage is a Class X Minus instrument designed for precise measurement applications, ideal for calibrating measuring equipment or directly gauging parts. It features a diameter of 0.1880 inches and a length of 2 inches, constructed from durable tool steel with a 4 Micro coating for enhanced performance.
  • Includes certificate of accuracy with NIST traceability
  • 4 Micro coated finish
  • Manufactured from tool steel
  • Suitable for calibration of measuring equipment or direct part gauging