Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.2530 inch Diameter, 2 inch Length
Catalog No. 5000531535
Encompass
This pin gage is designed for precise measurement and calibration tasks. It features a specific diameter and length, is constructed from durable tool steel, and has a 4 micro coated finish for enhanced performance and longevity. Each gage comes with a certificate of accuracy, ensuring traceability to NIST standards.
  • Made from tool steel
  • Features a 4 micro coated finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Diameter of 0.2530 inch
  • Tolerance of -0.00004 inch
  • Length of 2 inch
Catalog No. 50-005-31535 Supplier MSC Supplier No. 88825302
This pin gage is designed for precise measurement and calibration tasks. It features a specific diameter and length, is constructed from durable tool steel, and has a 4 micro coated finish for enhanced performance and longevity. Each gage comes with a certificate of accuracy, ensuring traceability to NIST standards.
  • Made from tool steel
  • Features a 4 micro coated finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Diameter of 0.2530 inch
  • Tolerance of -0.00004 inch
  • Length of 2 inch