Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.4995 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000535714
Encompass
This Class X Minus Pin Gage is designed for precise measurement and calibration in industrial applications. Crafted from durable tool steel, it features a 0.4995 inch diameter and a 2 inch length, ensuring accuracy and reliability for gaging parts or calibrating equipment.
  • Class X minus tolerance for high precision.
  • Manufactured from durable tool steel.
  • Features a 0.4995 inch diameter.
  • Measures 2 inches in length.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for calibrating measuring equipment or direct part gaging.
  • Provides a 4 micro coating/finish.
Catalog No. 50-005-35714 Supplier MSC Supplier No. 88849955
This Class X Minus Pin Gage is designed for precise measurement and calibration in industrial applications. Crafted from durable tool steel, it features a 0.4995 inch diameter and a 2 inch length, ensuring accuracy and reliability for gaging parts or calibrating equipment.
  • Class X minus tolerance for high precision.
  • Manufactured from durable tool steel.
  • Features a 0.4995 inch diameter.
  • Measures 2 inches in length.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for calibrating measuring equipment or direct part gaging.
  • Provides a 4 micro coating/finish.