Learn More
MSC Vermont Gage Pin Gage, Class X Minus, 0.5180 in Diameter
Catalog No. 5000510532
Encompass
This Class X Minus pin gage features a 0.5180 in diameter and a 2 in length. It is constructed from tool steel and has a 4 micro coating/finish. This gage is intended for use in the calibration of measuring equipment or for direct gaging of parts, and it comes with a Certificate of Accuracy with NIST Traceability.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 4 micro coating/finish.
  • Suitable for calibrating measuring equipment and direct part gaging.
Catalog No. 50-005-10532 Supplier MSC Supplier No. 88851803
This Class X Minus pin gage features a 0.5180 in diameter and a 2 in length. It is constructed from tool steel and has a 4 micro coating/finish. This gage is intended for use in the calibration of measuring equipment or for direct gaging of parts, and it comes with a Certificate of Accuracy with NIST Traceability.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 4 micro coating/finish.
  • Suitable for calibrating measuring equipment and direct part gaging.