Learn More
MSC Vermont Gage Pin Gage, Class X No-Go, 0.1390 in, 2 in
Catalog No. 5000516402
Encompass
This Class X No-Go (Minus) Pin Gage is a high-precision measurement tool designed to test the upper limit of bores or holes. It ensures that a manufactured part does not exceed the maximum allowable size, providing accurate verification of dimensions with a tight tolerance.
  • High-precision measurement tool with Class X designation.
  • Tight tolerance of 0.00004 ensures accuracy.
  • No-Go design tests the upper limit of bores or holes.
  • Ensures manufactured parts do not exceed maximum allowable size.
Catalog No. 50-005-16402 Supplier MSC Supplier No. 88813902
This Class X No-Go (Minus) Pin Gage is a high-precision measurement tool designed to test the upper limit of bores or holes. It ensures that a manufactured part does not exceed the maximum allowable size, providing accurate verification of dimensions with a tight tolerance.
  • High-precision measurement tool with Class X designation.
  • Tight tolerance of 0.00004 ensures accuracy.
  • No-Go design tests the upper limit of bores or holes.
  • Ensures manufactured parts do not exceed maximum allowable size.