Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.0710 in, 2 in
Catalog No. 5000531605
Encompass
This pin gage is a Class X Plus with a 0.0710 inch diameter and 2 inch length. It features a 4 Micro Coated finish, is made of tool steel, and includes a Certificate of Accuracy with NIST Traceability. It is suitable for calibrating measuring equipment or directly gaging parts.
  • Class X Plus pin gage
  • 0.0710 inch diameter
  • 2 inch length
  • 4 micro coated finish
  • Made from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment
  • Suitable for gaging parts directly
  • Country of origin: United States
Catalog No. 50-005-31605 Supplier MSC Supplier No. 88707104
This pin gage is a Class X Plus with a 0.0710 inch diameter and 2 inch length. It features a 4 Micro Coated finish, is made of tool steel, and includes a Certificate of Accuracy with NIST Traceability. It is suitable for calibrating measuring equipment or directly gaging parts.
  • Class X Plus pin gage
  • 0.0710 inch diameter
  • 2 inch length
  • 4 micro coated finish
  • Made from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment
  • Suitable for gaging parts directly
  • Country of origin: United States