Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.0770 in Diameter, 2 in Length
Catalog No. 5000515344
Encompass
This Class X Plus Pin Gage is designed for precise measurement applications, ideal for the calibration of measuring equipment or for directly gaging parts. Manufactured from durable tool steel, it offers reliability for critical inspection tasks.
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of +0.00004 inch
  • Manufactured from high-quality tool steel
  • Features a 4 micro finish
Catalog No. 50-005-15344 Supplier MSC Supplier No. 88707708
This Class X Plus Pin Gage is designed for precise measurement applications, ideal for the calibration of measuring equipment or for directly gaging parts. Manufactured from durable tool steel, it offers reliability for critical inspection tasks.
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of +0.00004 inch
  • Manufactured from high-quality tool steel
  • Features a 4 micro finish