Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.0800" Diameter, 2" Long
Catalog No. 5000534440
Encompass
This pin gage is designed for precise measurement applications, featuring a 0.0800 inch diameter and 2 inch length. It is constructed from durable tool steel with a 4 micro finish, ensuring accuracy and reliability for demanding tasks.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibrating measuring equipment or directly gaging parts
  • 0.0800 inch measurement diameter
  • 2 inch length
  • Class X Plus precision
  • 4 micro finish for smooth surface
  • Constructed from tool steel for durability
Catalog No. 50-005-34440 Supplier MSC Supplier No. 88708003
This pin gage is designed for precise measurement applications, featuring a 0.0800 inch diameter and 2 inch length. It is constructed from durable tool steel with a 4 micro finish, ensuring accuracy and reliability for demanding tasks.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibrating measuring equipment or directly gaging parts
  • 0.0800 inch measurement diameter
  • 2 inch length
  • Class X Plus precision
  • 4 micro finish for smooth surface
  • Constructed from tool steel for durability