Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.0870 in, 2 in
Catalog No. 5000534537
Encompass
This pin gage is a Class X Plus precision tool designed for accurate measurement. It features a diameter of 0.0870 inches and a length of 2 inches, and comes with a 4 micro coated finish.
  • Class X Plus for high precision
  • 0.0870 inch diameter
  • 2 inch length
  • 4 micro coated finish
Catalog No. 50-005-34537 Supplier MSC Supplier No. 88708706
This pin gage is a Class X Plus precision tool designed for accurate measurement. It features a diameter of 0.0870 inches and a length of 2 inches, and comes with a 4 micro coated finish.
  • Class X Plus for high precision
  • 0.0870 inch diameter
  • 2 inch length
  • 4 micro coated finish