Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1300 in Diameter, 2 in Length
Catalog No. 5000510320
Encompass
This pin gage is a Class X Plus instrument designed for precise measurement and calibration. It is constructed from durable tool steel and features a 4 Micro coating for enhanced performance. This gage is ideal for calibrating measuring equipment or directly gauging parts, and it includes a Certificate of Accuracy with NIST Traceability for assured quality.
  • Class X Plus for high accuracy applications
  • Constructed from durable tool steel
  • Features a 4 Micro coated finish
  • Suitable for calibrating measuring equipment
  • Can be used for direct part gauging
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-10320 Supplier MSC Supplier No. 88713003
This pin gage is a Class X Plus instrument designed for precise measurement and calibration. It is constructed from durable tool steel and features a 4 Micro coating for enhanced performance. This gage is ideal for calibrating measuring equipment or directly gauging parts, and it includes a Certificate of Accuracy with NIST Traceability for assured quality.
  • Class X Plus for high accuracy applications
  • Constructed from durable tool steel
  • Features a 4 Micro coated finish
  • Suitable for calibrating measuring equipment
  • Can be used for direct part gauging
  • Includes a certificate of accuracy with NIST traceability