Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1370 in, 2 in
Catalog No. 5000531078
Encompass
This pin gage is designed for the calibration of measuring equipment or for direct gaging of parts. It features a 0.1370 inch diameter and a 2 inch length, and is made from durable tool steel with a 4 Micro coating. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring precision for critical applications.
  • Designed for calibration of measuring equipment or direct gaging of parts.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel.
  • Features a 4 micro coating.
  • Precision diameter of 0.1370 "
  • Overall length of 2 inches.
  • Class X Plus tolerance.
Catalog No. 50-005-31078 Supplier MSC Supplier No. 88713706
This pin gage is designed for the calibration of measuring equipment or for direct gaging of parts. It features a 0.1370 inch diameter and a 2 inch length, and is made from durable tool steel with a 4 Micro coating. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring precision for critical applications.
  • Designed for calibration of measuring equipment or direct gaging of parts.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel.
  • Features a 4 micro coating.
  • Precision diameter of 0.1370 "
  • Overall length of 2 inches.
  • Class X Plus tolerance.