Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1700 in, 2 in
Catalog No. 5000534489
Encompass
This pin gage is a precision measuring tool designed for accurate calibration and direct gaging of parts. With a diameter of 0.1700 in and a length of 2 in, it is constructed from high-quality tool steel, ensuring durability and reliability for demanding applications. It comes with a Certificate of Accuracy with NIST Traceability for assured precision.
  • Designed for high-precision measurement applications.
  • Constructed from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for calibrating measuring equipment or gaging parts directly.
  • Features a 4 micro coating/finish for enhanced performance.
Catalog No. 50-005-34489 Supplier MSC Supplier No. 88717004
This pin gage is a precision measuring tool designed for accurate calibration and direct gaging of parts. With a diameter of 0.1700 in and a length of 2 in, it is constructed from high-quality tool steel, ensuring durability and reliability for demanding applications. It comes with a Certificate of Accuracy with NIST Traceability for assured precision.
  • Designed for high-precision measurement applications.
  • Constructed from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for calibrating measuring equipment or gaging parts directly.
  • Features a 4 micro coating/finish for enhanced performance.