Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1870 in Diameter, 2 in Length
Catalog No. 5000531144
Encompass
This Class X Plus Pin Gage features a 0.1870 inch diameter and a 2 inch length. It is constructed from tool steel and has a 4 micro coating, making it suitable for precise calibration of measuring equipment or direct gaging of parts.
  • Constructed from tool steel
  • Features a 0.1870 in diameter
  • Features a 2 in length
  • Includes a 4 micro coating
  • Comes with a certificate of accuracy with NIST traceability
  • Ideal for calibration of measuring equipment
  • Suitable for direct gaging of parts
Catalog No. 50-005-31144 Supplier MSC Supplier No. 88718705
This Class X Plus Pin Gage features a 0.1870 inch diameter and a 2 inch length. It is constructed from tool steel and has a 4 micro coating, making it suitable for precise calibration of measuring equipment or direct gaging of parts.
  • Constructed from tool steel
  • Features a 0.1870 in diameter
  • Features a 2 in length
  • Includes a 4 micro coating
  • Comes with a certificate of accuracy with NIST traceability
  • Ideal for calibration of measuring equipment
  • Suitable for direct gaging of parts