Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1965 in Dia
Catalog No. 5000510324
Encompass
This Class X Plus pin gage is designed for precise measurement and calibration. Made from durable tool steel, it features a 0.1965 inch diameter, a 2 inch length, and a 4 micro finish, ensuring high accuracy for critical applications.
  • Class X Plus accuracy
  • 0.1965 inch diameter
  • 2 inch length
  • Tool steel construction
  • 4 micro finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-10324 Supplier MSC Supplier No. 88719653
This Class X Plus pin gage is designed for precise measurement and calibration. Made from durable tool steel, it features a 0.1965 inch diameter, a 2 inch length, and a 4 micro finish, ensuring high accuracy for critical applications.
  • Class X Plus accuracy
  • 0.1965 inch diameter
  • 2 inch length
  • Tool steel construction
  • 4 micro finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability