Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.1990 in Diameter, 2 in Length
Catalog No. 5000534399
Encompass
This pin gage is designed for precise measurement and calibration tasks. It features a 0.1990 inch diameter and a 2 inch length, suitable for gaging parts directly or calibrating measuring equipment. The gage has a 4 Micro coated finish, ensuring durability and accuracy.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-34399 Supplier MSC Supplier No. 88719901
This pin gage is designed for precise measurement and calibration tasks. It features a 0.1990 inch diameter and a 2 inch length, suitable for gaging parts directly or calibrating measuring equipment. The gage has a 4 Micro coated finish, ensuring durability and accuracy.
  • Includes a certificate of accuracy with NIST traceability.