Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.2040 in, 2 in
Catalog No. 5000510996
Encompass
This pin gage is a Class X Plus instrument, meticulously crafted from tool steel, featuring a precise 0.2040 in diameter and a 2 in length. It is designed for accurate measurement applications, such as the calibration of measuring equipment or direct part gaging, ensuring reliable results.
  • Class X Plus precision
  • 0.2040 in diameter
  • 2 in length
  • 4 micro coated finish
  • Made from durable tool steel
  • Tolerance of 0.00004 in
  • Ideal for calibrating measuring equipment
  • Can be used for direct part gaging
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-10996 Supplier MSC Supplier No. 88720404
This pin gage is a Class X Plus instrument, meticulously crafted from tool steel, featuring a precise 0.2040 in diameter and a 2 in length. It is designed for accurate measurement applications, such as the calibration of measuring equipment or direct part gaging, ensuring reliable results.
  • Class X Plus precision
  • 0.2040 in diameter
  • 2 in length
  • 4 micro coated finish
  • Made from durable tool steel
  • Tolerance of 0.00004 in
  • Ideal for calibrating measuring equipment
  • Can be used for direct part gaging
  • Includes certificate of accuracy with NIST traceability