Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.2390 in Dia, 2 in Long
Catalog No. 5000530257
Encompass
This precision pin gage is designed for the calibration of measuring equipment or for direct part gauging. It features a 0.2390 inch diameter and a 2 inch length, crafted from durable tool steel with a 4 Micro coating for enhanced performance.
  • Designed for calibration of measuring equipment or to gage parts directly.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Features a 4 Micro coating/finish.
  • Class X Plus precision.
Catalog No. 50-005-30257 Supplier MSC Supplier No. 88723903
This precision pin gage is designed for the calibration of measuring equipment or for direct part gauging. It features a 0.2390 inch diameter and a 2 inch length, crafted from durable tool steel with a 4 Micro coating for enhanced performance.
  • Designed for calibration of measuring equipment or to gage parts directly.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Features a 4 Micro coating/finish.
  • Class X Plus precision.