Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.2430 in Diameter, Tool Steel, 2 in Length
Catalog No. 5000516197
Encompass
This Class X Plus pin gage is manufactured from tool steel and features a 0.2430 inch diameter and 2 inch length. It is designed for precise measurement applications, including the calibration of measuring equipment or direct part gaging, and comes with a certificate of accuracy.
  • Accurate and cost-effective measurement standards
  • Gauge tolerance of +0.00004 in.
  • Tool steel construction
  • 4 microinch finish
  • Includes certificate of accuracy with NIST traceability
  • ISO 17025 accredited and non-accredited calibration certificates are available for purchase
Catalog No. 50-005-16197 Supplier MSC Supplier No. 88724307
This Class X Plus pin gage is manufactured from tool steel and features a 0.2430 inch diameter and 2 inch length. It is designed for precise measurement applications, including the calibration of measuring equipment or direct part gaging, and comes with a certificate of accuracy.
  • Accurate and cost-effective measurement standards
  • Gauge tolerance of +0.00004 in.
  • Tool steel construction
  • 4 microinch finish
  • Includes certificate of accuracy with NIST traceability
  • ISO 17025 accredited and non-accredited calibration certificates are available for purchase