Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.2760 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000531223
Encompass
This pin gage is manufactured from tool steel, providing durability and stability for precise measurements. It is designed for accurate calibration of measuring equipment or for directly gaging parts, ensuring reliable performance in various applications.
  • Class X Plus for high precision
  • Diameter of 0.2760 inch
  • Length of 2 inch
  • Constructed from durable tool steel
  • Tolerance of 0.00004 inch
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for calibrating measuring equipment
  • Suitable for directly gaging parts
Catalog No. 50-005-31223 Supplier MSC Supplier No. 88727607
This pin gage is manufactured from tool steel, providing durability and stability for precise measurements. It is designed for accurate calibration of measuring equipment or for directly gaging parts, ensuring reliable performance in various applications.
  • Class X Plus for high precision
  • Diameter of 0.2760 inch
  • Length of 2 inch
  • Constructed from durable tool steel
  • Tolerance of 0.00004 inch
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for calibrating measuring equipment
  • Suitable for directly gaging parts