Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.3090 in Dia, 2 in Long
Catalog No. 5000534509
Encompass
This Class X Plus pin gage is designed for precise measurement and calibration tasks. It features a 0.3090 in diameter and a 2 in length, constructed from durable tool steel with a 4 Micro coating for enhanced performance and longevity. It is suitable for calibrating measuring equipment or directly gaging parts.
  • 0.3090 in diameter
  • 2 in length
  • 4 Micro coating
  • Made from tool steel
  • Tolerance of 0.00004 in
  • Ideal for calibrating measuring equipment or directly gaging parts
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-34509 Supplier MSC Supplier No. 88730908
This Class X Plus pin gage is designed for precise measurement and calibration tasks. It features a 0.3090 in diameter and a 2 in length, constructed from durable tool steel with a 4 Micro coating for enhanced performance and longevity. It is suitable for calibrating measuring equipment or directly gaging parts.
  • 0.3090 in diameter
  • 2 in length
  • 4 Micro coating
  • Made from tool steel
  • Tolerance of 0.00004 in
  • Ideal for calibrating measuring equipment or directly gaging parts
  • Includes a certificate of accuracy with NIST traceability