Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.3100 in Diameter, 2 in Length
Catalog No. 5000510322
Encompass
This Class X Plus Pin Gage is designed for precise calibration of measuring equipment or for direct gaging of parts. Manufactured from durable tool steel, it features a 0.3100 in diameter and a 2 in length, ensuring high accuracy with a tolerance of +0.00004 in. Each gage includes a Certificate of Accuracy with NIST Traceability, providing assurance of its precision and reliability.
  • Designed for calibration of measuring equipment or direct gaging of parts
  • Includes Certificate of Accuracy with NIST Traceability
  • Constructed from tool steel
  • Features a 4 micro coating/finish
  • Offers high precision with a +0.00004 in tolerance
Catalog No. 50-005-10322 Supplier MSC Supplier No. 88731005
This Class X Plus Pin Gage is designed for precise calibration of measuring equipment or for direct gaging of parts. Manufactured from durable tool steel, it features a 0.3100 in diameter and a 2 in length, ensuring high accuracy with a tolerance of +0.00004 in. Each gage includes a Certificate of Accuracy with NIST Traceability, providing assurance of its precision and reliability.
  • Designed for calibration of measuring equipment or direct gaging of parts
  • Includes Certificate of Accuracy with NIST Traceability
  • Constructed from tool steel
  • Features a 4 micro coating/finish
  • Offers high precision with a +0.00004 in tolerance