Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.3945 in, 2 in, 4 Micro Coated
Catalog No. 5000530261
Encompass
This Class X Plus Pin Gage has a diameter of 0.3945 inch and a length of 2 " It features a 4 Micro coating and is used for calibrating measuring equipment or directly gaging parts.
  • Includes certificate of accuracy with NIST traceability
  • 4 Micro coated
  • Made from tool steel
  • Suitable for calibration of measuring equipment or to gage parts directly
Catalog No. 50-005-30261 Supplier MSC Supplier No. 88739453
This Class X Plus Pin Gage has a diameter of 0.3945 inch and a length of 2 " It features a 4 Micro coating and is used for calibrating measuring equipment or directly gaging parts.
  • Includes certificate of accuracy with NIST traceability
  • 4 Micro coated
  • Made from tool steel
  • Suitable for calibration of measuring equipment or to gage parts directly