Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.4000" Dia, 2" Long
Catalog No. 5000515877
Encompass
This Class X Plus Pin Gage features a diameter of 0.4000" and a length of 2". It is designed for the precise calibration of measuring equipment or for directly gaging parts, offering reliable performance with its 4 Micro Coated finish.
  • Features a 4 Micro Coated finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for the calibration of measuring equipment.
  • Can be used to gage parts directly.
Catalog No. 50-005-15877 Supplier MSC Supplier No. 88740006
This Class X Plus Pin Gage features a diameter of 0.4000" and a length of 2". It is designed for the precise calibration of measuring equipment or for directly gaging parts, offering reliable performance with its 4 Micro Coated finish.
  • Features a 4 Micro Coated finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for the calibration of measuring equipment.
  • Can be used to gage parts directly.