Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.4230 in, 2 in, Tool Steel
Catalog No. 5000529034
Encompass
This pin gage is designed for precise measurement and calibration in demanding applications. Manufactured from durable tool steel, it offers exceptional accuracy for calibrating measuring equipment or directly gaging parts. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Designed for calibration of measuring equipment or to gage parts directly.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Manufactured from robust tool steel for durability.
  • Features a 4 micro coating/finish.
  • Provides a precise diameter of 0.4230 "
  • Offers a tight tolerance of 0.00004 "
  • Measures 2 inch in length.
  • Class X plus precision.
Catalog No. 50-005-29034 Supplier MSC Supplier No. 88742309
This pin gage is designed for precise measurement and calibration in demanding applications. Manufactured from durable tool steel, it offers exceptional accuracy for calibrating measuring equipment or directly gaging parts. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Designed for calibration of measuring equipment or to gage parts directly.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Manufactured from robust tool steel for durability.
  • Features a 4 micro coating/finish.
  • Provides a precise diameter of 0.4230 "
  • Offers a tight tolerance of 0.00004 "
  • Measures 2 inch in length.
  • Class X plus precision.