Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.4240 in Diameter, 2 in Length
Catalog No. 5000515876
Encompass
This pin gage is a Class X Plus tool, featuring a diameter of 0.4240 in and a length of 2 in. Manufactured from tool steel with a 4 micro coating, it is designed for the calibration of measuring equipment or for direct part gaging.
  • Class X Plus pin gage
  • 0.4240 in diameter
  • 2 in length
  • 0.00004 in tolerance
  • Tool steel construction
  • 4 micro coating
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-15876 Supplier MSC Supplier No. 88742408
This pin gage is a Class X Plus tool, featuring a diameter of 0.4240 in and a length of 2 in. Manufactured from tool steel with a 4 micro coating, it is designed for the calibration of measuring equipment or for direct part gaging.
  • Class X Plus pin gage
  • 0.4240 in diameter
  • 2 in length
  • 0.00004 in tolerance
  • Tool steel construction
  • 4 micro coating
  • Includes certificate of accuracy with NIST traceability