Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.4380 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000527557
Encompass
This Class X Plus Pin Gage features a 0.4380 in diameter and a 2 in length, designed for precise measurement applications. It is constructed from tool steel and features a 4 micro coated finish, ensuring durability and accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for calibration of measuring equipment.
  • Can be used to gage parts directly.
  • Constructed from tool steel.
  • Features a 4 micro coated finish.
  • Tolerance of 0.00004 in.
Catalog No. 50-005-27557 Supplier MSC Supplier No. 88743802
This Class X Plus Pin Gage features a 0.4380 in diameter and a 2 in length, designed for precise measurement applications. It is constructed from tool steel and features a 4 micro coated finish, ensuring durability and accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for calibration of measuring equipment.
  • Can be used to gage parts directly.
  • Constructed from tool steel.
  • Features a 4 micro coated finish.
  • Tolerance of 0.00004 in.