Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.4400 in, 2 in
Catalog No. 5000536701
Encompass
This pin gage is a Class X Go gage, utilized as a master for calibrating equipment or for directly gaging parts. It serves as an accurate, cost-effective measurement standard for both shop floor and metrology laboratory applications.
  • Plus tolerance for go gages
  • Made from tool steel 60/65 Rc
  • 2 in long
  • Finish of 4 microinch or better
  • Round within 1/2 the diameter tolerance
  • Includes a certificate of accuracy
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased
Catalog No. 50-005-36701 Supplier MSC Supplier No. 88744008
This pin gage is a Class X Go gage, utilized as a master for calibrating equipment or for directly gaging parts. It serves as an accurate, cost-effective measurement standard for both shop floor and metrology laboratory applications.
  • Plus tolerance for go gages
  • Made from tool steel 60/65 Rc
  • 2 in long
  • Finish of 4 microinch or better
  • Round within 1/2 the diameter tolerance
  • Includes a certificate of accuracy
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased