Learn More
MSC Vermont Gage Pin Gage, Class X Plus, 0.6000 in, 2 in, Tool Steel
Catalog No. 5000535430
Encompass
This pin gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. It features a Class X Plus tolerance, ensuring high accuracy for critical tasks.
  • Manufactured from tool steel for durability.
  • Features a 0.6000 inch diameter and 2 inch length for versatile applications.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Designed with a 4 micro coating/finish for enhanced performance.
Catalog No. 50-005-35430 Supplier MSC Supplier No. 88760004
This pin gage is designed for precise measurement applications, ideal for calibrating measuring equipment or directly gaging parts. It features a Class X Plus tolerance, ensuring high accuracy for critical tasks.
  • Manufactured from tool steel for durability.
  • Features a 0.6000 inch diameter and 2 inch length for versatile applications.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Designed with a 4 micro coating/finish for enhanced performance.