Learn More
MSC Vermont Gage Pin Gage, Class X Plus, Steel, 0.3140 inch Diameter, 2 inch Long
Catalog No. 5000511824
Encompass
This Class X Plus pin gage is designed for precise measurement and calibration tasks. Manufactured from durable tool steel, it features a diameter of 0.3140 inches and a length of 2 inches. It is suitable for both direct part gaging and calibrating measuring equipment in various laboratory and industrial settings.
  • Made from durable tool steel.
  • Features a precise 0.3140 inch diameter.
  • Provides a length of 2 inches.
  • Offers a tight tolerance of +0.00004 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Ensures a smooth 4 microinch finish.
  • Ideal for calibrating equipment or directly gaging parts.
Catalog No. 50-005-11824 Supplier MSC Supplier No. 88731401
This Class X Plus pin gage is designed for precise measurement and calibration tasks. Manufactured from durable tool steel, it features a diameter of 0.3140 inches and a length of 2 inches. It is suitable for both direct part gaging and calibrating measuring equipment in various laboratory and industrial settings.
  • Made from durable tool steel.
  • Features a precise 0.3140 inch diameter.
  • Provides a length of 2 inches.
  • Offers a tight tolerance of +0.00004 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Ensures a smooth 4 microinch finish.
  • Ideal for calibrating equipment or directly gaging parts.