Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 0.3045 Diameter, NoGo (Minus)
Catalog No. 5000531368
Encompass
This pin gage is designed for precise measurement of hole sizes. It features a 0.3045 inch diameter and is a Class ZZ, NoGo (Minus) type, crafted from durable tool steel with a hardness of Rc 60/65. It measures hole sizes within a tolerance of +.0000/-.0002 inch and includes a Certificate of Accuracy with NIST Traceability.
  • Precisely measures hole sizes.
  • Class ZZ tolerance.
  • 0.3045 inch diameter.
  • NoGo (minus) type for specific gauging needs.
  • Constructed from tool steel with Rc 60/65 hardness for durability.
  • Overall length of 2 inches.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-31368 Supplier MSC Supplier No. 89030456
This pin gage is designed for precise measurement of hole sizes. It features a 0.3045 inch diameter and is a Class ZZ, NoGo (Minus) type, crafted from durable tool steel with a hardness of Rc 60/65. It measures hole sizes within a tolerance of +.0000/-.0002 inch and includes a Certificate of Accuracy with NIST Traceability.
  • Precisely measures hole sizes.
  • Class ZZ tolerance.
  • 0.3045 inch diameter.
  • NoGo (minus) type for specific gauging needs.
  • Constructed from tool steel with Rc 60/65 hardness for durability.
  • Overall length of 2 inches.
  • Includes a certificate of accuracy with NIST traceability.