Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 10.07 mm, Minus Tolerance, Steel
Catalog No. 5000528459
Encompass
This Vermont Gage pin gage is a Class ZZ, steel gage designed for precise measurement. With a diameter of 10.07 mm and a minus tolerance of 0.00508 mm, it is suitable for setting micrometers, verniers, and other measuring devices, particularly for Nogo inspection.
  • Designed for setting micrometers, for Nogo inspection, verniers and other measuring devices.
  • Class ZZ for precise gaging.
  • 10.07 mm diameter for specific measurement requirements.
  • Features a minus direction of tolerance.
  • Constructed from durable steel.
  • Reversible style for extended use.
  • Metric system of measurement.
  • Tolerance of 0.00508 mm.
Catalog No. 50-005-28459 Supplier MSC Supplier No. 40437527
This Vermont Gage pin gage is a Class ZZ, steel gage designed for precise measurement. With a diameter of 10.07 mm and a minus tolerance of 0.00508 mm, it is suitable for setting micrometers, verniers, and other measuring devices, particularly for Nogo inspection.
  • Designed for setting micrometers, for Nogo inspection, verniers and other measuring devices.
  • Class ZZ for precise gaging.
  • 10.07 mm diameter for specific measurement requirements.
  • Features a minus direction of tolerance.
  • Constructed from durable steel.
  • Reversible style for extended use.
  • Metric system of measurement.
  • Tolerance of 0.00508 mm.