Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 18.89 mm, Plus, Tool Steel
Catalog No. 5000527683
Encompass
This Vermont Gage pin gage is designed for precise measurement of hole sizes within a tolerance of +.005/-.000. It features an 18.89 mm diameter and is made from durable tool steel with an Rc 60/65 hardness. The gage is of Class ZZ and comes with a certificate of accuracy traceable to NIST.
  • Made from tool steel with Rc 60/65 hardness
  • Measures hole sizes within a tolerance of +.005/-.000 inch
  • Includes certificate of accuracy with NIST traceability
  • Overall length is 2 in
  • Features a Go (Plus) type
  • Class ZZ for precision
  • Finish of steel with 10 microfinish or better
Catalog No. 50-005-27683 Supplier MSC Supplier No. 68109941
This Vermont Gage pin gage is designed for precise measurement of hole sizes within a tolerance of +.005/-.000. It features an 18.89 mm diameter and is made from durable tool steel with an Rc 60/65 hardness. The gage is of Class ZZ and comes with a certificate of accuracy traceable to NIST.
  • Made from tool steel with Rc 60/65 hardness
  • Measures hole sizes within a tolerance of +.005/-.000 inch
  • Includes certificate of accuracy with NIST traceability
  • Overall length is 2 in
  • Features a Go (Plus) type
  • Class ZZ for precision
  • Finish of steel with 10 microfinish or better