Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 21.17 mm, Tool Steel
Catalog No. 5000516446
Encompass
This Vermont Gage Class ZZ Minus Pin Gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell hardness of Rc 60/65, it ensures accuracy within a tolerance of +.000/-.005. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Measures hole sizes within +.000/-.005 in tolerance.
  • Includes certificate of accuracy with NIST traceability.
  • Features a steel with 10 microfinish or better.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Overall length is 2 in.
Catalog No. 50-005-16446 Supplier MSC Supplier No. 40448623
This Vermont Gage Class ZZ Minus Pin Gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell hardness of Rc 60/65, it ensures accuracy within a tolerance of +.000/-.005. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Measures hole sizes within +.000/-.005 in tolerance.
  • Includes certificate of accuracy with NIST traceability.
  • Features a steel with 10 microfinish or better.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Overall length is 2 in.