Learn More
MSC VERMONT GAGE Pin Gage, Class ZZ, 22.1 mm, Steel, Plus Tolerance
Catalog No. 5000531871
Encompass
This pin gage is designed for precise measurement applications, featuring a Class ZZ tolerance with a plus direction. Constructed from durable steel, it has a 22.1 mm size and is reversible for extended utility, utilizing a metric system of measurement.
  • Class ZZ tolerance
  • Plus tolerance direction
  • Constructed from steel
  • 22.1 mm size
  • Reversible style
  • Metric system of measurement
Catalog No. 50-005-31871 Supplier MSC Supplier No. 68113158
This pin gage is designed for precise measurement applications, featuring a Class ZZ tolerance with a plus direction. Constructed from durable steel, it has a 22.1 mm size and is reversible for extended utility, utilizing a metric system of measurement.
  • Class ZZ tolerance
  • Plus tolerance direction
  • Constructed from steel
  • 22.1 mm size
  • Reversible style
  • Metric system of measurement